To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Membrane protein
- Clonality:Polyclonal
- Gene ID:43740571
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Antigen symbol:M
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.02 mg
- Form:Liquid
- Antigen synonyms:SARS Matrix protein|SARS-CoV-2/SARS-CoV Matrix Antibody: SARS-CoV-2 Matrix protein|Membrane protein|M protein|E1 glycoprotein
- Modification:None
- Storage buffer:SARS-CoV-2 (COVID-19) Membrane antibody is supplied in PBS containing 0.02% sodium azide.
- Concentration:1 mg/mL
- Shipping temperature:Blue ice
- Immunogen:Anti-SARS-CoV-2 (COVID-19) Membrane antibody (biotin) 9157 was raised against a peptide corresponding to 13 amino acids near the center of SARS-CoV-2 (COVID-19) Membrane protein. The immunogen is located between 130-180 amino acids of the SARS-CoV-2 (COVID-19) Membrane protein.
- Purification:SARS-CoV-2 (COVID-19) Membrane antibody is affinity chromatography.
- Packaging:Vial
Spécifications
A propos de cet article
SARS-CoV-2 (COVID-19) Membrane antibody can be stored at 4 °C for three months and −20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type: Primary
Antigen: Membrane Protein
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus