Anti-KREMEN2 Rabbit Polyclonal Antibody
Catalog # 76754-886
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Kremen protein 2
- Antigen symbol:KREMEN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Size:400 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:79412
- Antigen synonyms:Kremen protein 2|KRM2|KREMEN2|Dickkopf receptor 2|Kringle-containing protein marking the eye and the nose|Kringle domain-containing transmembrane protein 2
- Modification:None
- Storage buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Storage temperature:Store at 4 °C for three months and −20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:This KREMEN2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 248-276 amino acids from the Central region of human KREMEN2.
- Purification:This antibody is purified through a rotein A column, followed by peptide affinity purification.
- Cat. no.:76754-886
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: KREMEN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human