Order Entry
ContactUsLinkComponent
Lifeasible Bacillus thuringiensis Recombinant Cry1Ac from (E. coli)
Lifeasible Bacillus thuringiensis Recombinant Cry1Ac from (E. coli)
# de catalogue 76683-052
Fournisseur:  Creative Biomart
Numéro CAS:  
Lifeasible Bacillus thuringiensis Recombinant Cry1Ac from (E. coli)
# de catalogue 76683-052
Fournisseur:  Creative Biomart
Numéro CAS:  

Spécifications

  • Animal-free:
    Y
  • Packaging:
    Vial
  • Protein Function:
    Insecticidal protein
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bacillus thuringiensis
  • Conjugation:
    Unconjugated
  • Size:
    1 mg
  • Storage conditions:
    –80 °C
  • Protein/peptide name:
    Cry1Ac
  • Purity:
    85%
  • Molecular weight:
    130.7 kDa
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    –80 °C
  • Cat. no.:
    76683-052

Spécifications

A propos de cet article

Cry1Ac is a parasporal crystal or insecticidal crystal protein or delta endotoxin produced by Bt, which has insecticidal activity against a variety of pests. Cry1Ac protoxin is activated by the intestinal juice of the target pest to obtain an activated toxin fragment, which binds to the midgut epithelial cells of the pest to form holes, leading to intestinal paralysis and eventually death due to sepsis.

Sequence:
MDNNPNINECIPYNCLSNPEVEVLGGERIETGYTPIDISLSLTQFLLSEFVPGAGFVLGLVDIIWGIFGPSQWDAFLVQIEQLINQRIEEFARNQAISRLEGLSNLYQIYAESFREWEADPTNPALREEMRIQFNDMNSALTTAIPLFAVQNYQVPLLSVYVQAANLHLSVLRDVSVFGQRWGFDAATINSRYNDLTRLIGNYTDYAVRWYNTGLERVWGPDSRDWVRYNQFRRELTLTVLDIVALFPNYDSRRYPIRTVSQLTREIYTNPVLENFDGSFRGSAQGIERSIRSPHLMDILNSITIYTDAHRGYYYWSGHQIMASPVGFSGPEFTFPLYGTMGNAAPQQRIVAQLGQGVYRTLSSTLYRRPFNIGINNQQLSVLDGTEFAYGTSSNLPSAVYRKSGTVDSLDEIPPQNNNVPPRQGFSHRLSHVSMFRSGFSNSSVSIIRAPMFSWIHRSAEFNNIIASDSITQIPAVKGNFLFNGSVISGPGFTGGDLVRLNSSGNNIQNRGYIEVPIHFPSTSTRYRVRVRYASVTPIHLNVNWGNSSIFSNTVPATATSLDNLQSSDFGYFESANAFTSSLGNIVGVRNFSGTAGVIIDRFEFIPVTATLEAESDLERAQKAVNALFTSSNQIGLKTDVTDYHIDRVSNLVECLSDEFCLDEKKELSEKVKHAKRLSDERNLLQDPNFRGINRQLDRGWRGSTDITIQGGDDVFKENYVTLLGTFDECYPTYLYQKIDESKLKAYTRYQLRGYIEDSQDLEIYLIRYNAKHETVNVPGTGSLWPLSAPSPIGKCAHHSHHFSLDIDVGCTDLNEDLGVWVIFKIKTQDGHARLGNLEFLEEKPLVGEALARVKRAEKKWRDKREKLEWETNIVYKEAKESVDALFVNSQYDRLQADTNIAMIHAADKRVHSIREAYLPELSVIPGVNAAIFEELEGRIFTAFSLYDARNVIKNGDFNNGLSCWNVKGHVDVEEQNNHRSVLVVPEWEAEVSQEVRVCPGRGYILRVTAYKEGYGEGCVTIHEIENNTDELKFSNCVEEEVYPNNTVTCNDYTATQEEYEGTYTSRNRGYDGAYESNSSVPADYASAYEEKAYTDGRRDNPCESNRGYGDYTPLPAGYVTKELEYFPETDKVWIEIGETEGTFIVDSVELLLMEE

Delivery: Dry ice.
Customers Who Bought This Also Bought