Lifeasible Bacillus thuringiensis Recombinant Cry2Ab from (E. coli)
# de catalogue 76683-092
Fournisseur: Creative Biomart
Numéro CAS:
Spécifications
- Animal-free:Y
- Packaging:Vial
- Protein Function:Insecticidal protein
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Bacillus thuringiensis
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–80 °C
- Protein/peptide name:Cry2Ab
- Purity:80%
- Molecular weight:75 kDa
- Sequence:MDNSVLNSGRTTICDAYNVAAHDPFSFQHKSLDTVQKEWTEWKKNNHSLYLDPIVGTVASFLLKKVGSLVGKRILSELRNLIFPSGSTNLMQDILRETEKFLNQRLNTDTLARVNAELTGLQANVEEFNRQVDNFLNPNRNAVPLSITSSVNTMQQLFLNRLPQFQMQGYQLLLLPLFAQAANLHLSFIRDVILNADEWGISAATLRTYRDYLKNYTRDYSNYCINTYQSAFKGLNTRLHDMLEFRTYMFLNVFEYVSIWSLFKYQSLLVSSGANLYASGSGPQQTQSFTSQDWPFLYSLFQVNSNYVLNGFSGARLSNTFPNIVGLPGSTTTHALLAARVNYSGGISSGDIGASPFNQNFNCSTFLPPLLTPFVRSWLDSGSDREGVATVTNWQTESFETTLGLRSGAFTARGNSNYFPDYFIRNISGVPLVVRNEDLRRPLHYNEIRNIASPSGTPGGARAYMVSVHNRKNNIHAVHENGSMIHLAPNDYTGFTISPIHATQVNNQTRTFISEKFGNQGDSLRFEQNNTTARYTLRGNGNSYNLYLRVSSIGNSTIRVTINGRVYTATNVNTTTNNDGVNDNGARFSDINIGNVVASSNSDVPLDINVTLNSGTQFDLMNIMLVPTNISPLY
- Concentration:1 mg/ml
- Shipping temperature:–80 °C
- Cat. no.:76683-092
Spécifications
A propos de cet article
Promotes colloidosmotic lysis by binding to the midgut epithelial cells of lepidopteran (Manduca sexta) larvae.
Delivery: Dry ice.
Customers Who Bought This Also Bought