Lifeasible Bacillus thuringiensis Recombinant Cry1Da2 from (E. coli)
# de catalogue 76683-080
Fournisseur: Creative Biomart
Numéro CAS:
Spécifications
- Animal-free:Y
- Packaging:Vial
- Protein Function:Insecticidal protein
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Bacillus thuringiensis
- Conjugation:Unconjugated
- Size:1 mg
- Storage conditions:–80 °C
- Protein/peptide name:Cry1Da2
- Purity:80%
- Molecular weight:132.5 kDa
- Sequence:MENNIQNQCVPYNCLNNPEVEILNEERSTGRLPLDISLSLTRFLLSEFVPGVGVAFGLFDLIWGFITPSDWSLFLLQIEQLIEQRIETLERNRAITTLRGLADSYEIYIEALREWEANPNNAQLREDVRIRFANTDDALITAINNFTLTSFEIPLLSVYVQAANLHLSLLRDAVSFGQGWGLDIATVNNHYNRLINLIHRYTKHCLDTYNQGLENLRGTNTRQWARFNQFRRDLTLTVLDIVALFPNYDVRTYPIQTSSQLTREIYTSSVIEDSPVSANIPNGFNRAEFGVRPPHLMDFMNSLFVTAETVRSQTVWGGHLVSSRNTAGNRINFPSYGVFNPGGAIWIADEDPRPFYRTLSDPVFVRGGFGNPHYVLGLRGVAFQQTGTNHTRTFRNSGTIDSLDEIPPQDNSGAPWNDYSHVLNHVTFVRWPGEISGSDSWRAPMFSWTHRSATPTNTIDPERITQIPLVKAHTLQSGTTVVRGPGFTGGDILRRTSGGPFAYTIVNINGQLPQRYRARIRYASTTNLRIYVTVAGERIFAGQFNKTMDTGDPLTFQSFSYATINTAFTFPMSQSSFTVGADTFSSGNEVYIDRFELIPVTATFE
- Concentration:1 mg/ml
- Shipping temperature:–80 °C
- Cat. no.:76683-080
Spécifications
A propos de cet article
Cry1Da2 is a parasporal crystal or insecticidal crystal protein or delta endotoxin produced by Bt, which has insecticidal activity against a variety of pests. The Cry1Da2 protoxin is activated by the intestinal juice of the target pest to obtain an activated toxin fragment, which binds to the midgut epithelial cells of the pest to form holes, leading to intestinal paralysis and eventually death due to sepsis.
Delivery: Dry ice.
Customers Who Bought This Also Bought