TTLL4 Peptide
# de catalogue 77128-558
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:9654
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Dog
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:OTTHUMP00000164099|tubulin tyrosine ligase-like family|TTLL4|member 4|KIAA0173
- Protein/peptide name:TTLL4
- Formulation:Lyophilized. TTLL4 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought