Hdac1 Peptide
# de catalogue 77132-794
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:433759
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Mouse
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:Hdac1-ps|RPD3|HD1|Histone deacetylase 1|MommeD5
- Protein/peptide name:Hdac1
- Formulation:Lyophilized. Hdac1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought