NPC2 Peptide
# de catalogue 77119-964
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:10577
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:Human epididymis-specific protein 1|EDDM1|HE1|Epididymal secretory protein E1|He1
- Protein/peptide name:NPC2
- Formulation:Lyophilized. NPC2 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77119-964
Spécifications
A propos de cet article
Customers Who Bought This Also Bought