Anti-null Recombinant Antibody [Clone: SAIC-19D-1F4]
# de catalogue 76629-570
Fournisseur: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:SAIC-19D-1F4
- Gene ID:3408
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:null
- Conjugation:Unconjugated
- Size:0.2 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Growth factor receptor-bound protein 2|ASH
- Modification:Unmodified
- Storage buffer:PBS with 0.02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide 'FGNDVQHFK', derived from the Growth factor receptor-bound protein 2, conjugated to KLH.
- Tested applications:immunoMRM
- Cat. no.:76629-570
- Packaging:Vial
Spécifications
A propos de cet article
Type: Primary
Antigen: null
Clonality: Recombinant
Clone: SAIC-19D-1F4
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human
Customers Who Bought This Also Bought