Eisco Porcelain Spot Plates
Fournisseur: Wards
Certificats
A propos de cet article
Spot plates made of high quality glazed porcelain which can be cleaned and reused.
- Spot plate made of high quality porcelain glazed on the top and sides
- Wells measure 22 mm in diameter
- Can be cleaned and reused
- Perfect for using with liquid or small solid samples
Plate is glazed on top and sides and can be washed and reused. Useful for comparing results among students in a lab, or in classroom demonstrations.
Customers Who Bought This Also Bought