VWR® Max Protection High ESD Cleanroom Lab Coats
Supplier: VWR International
Certificates
About this item
Cleanroom ESD lab coats are available with either a front snap closure or a front zipper closure.
- Lab coat featuring high ESD properties
- Lightweight comfort fit
- Snap or zipper closure options available
VWR® Max Protection high ESD cleanroom lab coats feature a collar, three pockets, elastic wrists, and serged seams. Garments are made from 63 gSF, and are packaged 25 per case.
Specifications
- Catalog No:
- 76919-412
- 76919-400
- 76919-394
- 76919-404
- 76919-392
- 76919-390
- 76919-414
- 76919-402
- 76919-410
- 76919-406
- 76919-396
- 76919-398
- 76919-408
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD