Fischertechnik TXT 4.0 Competition Add-On Pack
Catalog # 470332-600
Supplier: STUDICA INC.
Specifications
- Description:TXT 4.0 competition add-on pack
- Cat. no.:470332-600
Specifications
About this item
Enhance your TXT 4.0 Base Set robot for competitions.
- Pair with the TXT 4.0 Base set
- Teaching Topics Include: robot competitions, project work and workshops in robotics
- Suitable for Middle/High School students
- Downloadable teaching and activity guides
The Robotics ADD-ON Competition is designed for schools, universities and other educational institutions that want to develop or improve their models for robotics competitions for their students. With this set, models can be enhanced, and new features can be added, making this kit the perfect addition for competitions around the world. The set includes the new RGB guest sensor, a combo sensor (gyroscope, acceleration and compass), an ultrasonic sensor, two more powerful motors, and track links and caterpillar pads for the undercarriage of a tracked robot - ideal for building competitive driving robots.
Ordering information: Required but available separately: TXT 4.0 Base set.
Customers Who Bought This Also Bought