Nalgene™ Safety Waste Funnels, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
HDPE funnels with attached lid, polypropylene closure/funnel adapter, screen insert, polypropylene/PTFE 0.45µm vent plug, and filter. For use with Thermo Scientific™ Nalgene™ Safety Waste Funnel Systems.
- Funnels feature the Easy Snap Safety Latch to minimize spillage if accidentally tipped over and a larger diameter funnel for easy pouring
- The vent system helps reduce fumes and splashing while the removable neck screen traps large particles, like stir bars
- Designed for solvent, chemical and biological waste, it is recommended for use with Nalgene containers
Temporarily contain waste on the benchtop or under the lab hood using Thermo Scientific™ Nalgene™ Safety Waste Funnel Systems.
Customers Who Bought This Also Bought