Labconco® Heated Product Shelf Chambers with Valves
Fournisseur: Labconco
New Product
Certificats
A propos de cet article
Heated product shelf chambers with valves.
- Includes 12" dia. acrylic cylinder and base, 10" dia. removable shelves, and temperature probes
- Controlled heat settings−20 to 50 °C (−4 to 140 °F)
- Shelves do not cool
- Has 8" dia. opening and cap for 3" dia. attachment port
Customers Who Bought This Also Bought