Eisco Rectangular Retort Bases with Rubber Strips, Steel
Fournisseur: Wards
Certificats
A propos de cet article
Premium Retort Bases with no-slip rubber strips and tapped hole made of powder-coated pressed steel.
- Rectangular retort bases made of corrosion resistant, powder coated pressed steel
- Rubber strips on sort ends prevent slipping and protect surfaces
- Sturdy construction provides superb stability for experimental setups
- Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-weighted Retort Stands are made of corrosion resistant, powder coated pressed steel, and feature rubber strips at the short ends prevent slipping and protect surfaces. With a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.
Customers Who Bought This Also Bought