TETPOR AIR DEMI Capsules
Fournisseur: PARKER HANNIFIN TE
Certificats
A propos de cet article
TETPOR AIR sterilization filter capsules offer exceptional filtration performance while providing the highest levels of biosecurity throughout the process industry.
- Assured biosecurity with absolute rated filtration
- High flow rates with low pressure drops
- High voids volume PTFE membrane
- Unique prefilter layer
- Silicone seal material
Operating at ambient temperature conditions, TETPOR AIR filter capsules provide a cost-effective filtration solution. A polypropylene prefilter layer extends service life in heavily contaminated environments.
TETPOR AIR filter capsules also utilize a well-proven, inherently hydrophobic expanded PTFE membrane validated as sterilizing grade in liquid in accordance with ASTM F838-05. This ensures the removal of all airborne bacteria, viruses and bacteriophage.
Customers Who Bought This Also Bought