Laboratory Jacks, Steel
Fournisseur: Wards
Certificats
A propos de cet article
Height-adjustable lightweight lab jacks offer strong, stable support for laboratory apparatus.
- Adjustable and reversible Lab Jacks with screw-operated mechanism
- Made of powder coated lightweight steel resistant to many chemicals
- Single piece base provides enhanced torsional rigidity for stabilization under load
- Can be autoclaved and chemically cleaned
Strong, stable support for laboratory apparatus. The lab jack has a screw-operated scissor action mechanism for easy, fluid movement of platform - with and without load - and precise height adjustments. For physical, chemical, biological experiments and other uses.
Customers Who Bought This Also Bought