Coverslips
Fournisseur: Mortech Manufacturing
New Product
Certificats
A propos de cet article
Mopec coverslips are made with borosilicate glass.
- Borosilicate glass, #1 thickness, 12 mm in diameter
- Will not stick or auto-fluoresce
- Clean, uniform surface
- Ideal for automated coverslipping
Available in 1 or 2 ounce volumes.
Packaging: The coverslips are packaged 10 boxes per case, with 100 or 200 pieces per box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.