Eisco Woulff Gas Wash Bottles
Fournisseur: Wards
Certificats
A propos de cet article
Woulff gas wash bottles with two necks for preparing gases.
- Made of mechanically strong and chemical resistant Borosilicate 3.3 glass
- Two glass necks with ground sockets
- Used to remove impurities from gases, trapping particles and droplets, and for pressure equalization
- Autoclavable at 121 °C
Woulff gas wash bottles made from high quality, heavy duty Borosilicate 3.3 glass. Ground glass joints allow for easy sealing.
Customers Who Bought This Also Bought