Blank for Calibration Kit, A30
# de catalogue 76016-668
Fournisseur: FIREFLYSCI, INC.
Spécifications
- Description:Blank for calibration standards
- Cat. no.:76016-668
Spécifications
A propos de cet article
Blank for use in UV/VIS calibration kits.
- Anodized aluminum construction
- External dimensions: 12.5L×12.5W×45H mm
Customers Who Bought This Also Bought