Cuvette Mounts (For Lightpaths Below 5 mm), A20
Fournisseur: FIREFLYSCI, INC.
Certificats
A propos de cet article
Anodized aluminum mounts are available with or without extended spring.
- Anodized aluminum construction
- External dimensions: 12.5L×12.5W×52H mm
Customers Who Bought This Also Bought