Cuvette Rack, Type P66
# de catalogue 76016-654
Fournisseur: FIREFLYSCI, INC.
Spécifications
- Description:Polyethylene cuvette rack
- Cat. no.:76016-654
Spécifications
A propos de cet article
Cell holder rack for 12 standard size cuvettes.
- 12-hole cell rack
- Designed to hold twelve (12) 10 mm pathlength cells
Rack is manufactured from polyethylene to ensure that windows cannot be scratched or damaged when being placed in the rack.
Customers Who Bought This Also Bought