ContactUsLinkComponent
Control Slides, Tissue Microarray, Normal Human Tissue, 16 Tissue Types
Control Slides, Tissue Microarray, Normal Human Tissue, 16 Tissue Types
# de catalogue 76948-262
Fournisseur:  Mortech Manufacturing
New Product
CONTROL SLIDES HMN MICROARRAY 16/15 CS25
Control Slides, Tissue Microarray, Normal Human Tissue, 16 Tissue Types
# de catalogue 76948-262
Fournisseur:  Mortech Manufacturing
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Mopec Control slides, tissue microarray, normal human tissue, 16 tissue types
Spécifications
A propos de cet article
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 μm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Tissue type: chosen from Liver, Lung, Kidney, Colon, Tonsil, Skin, Prostate, Breast, Lymph Node, Uterus, Testis, Placenta, Thyroid, Stomach, and Pancreas.
Ordering information: Case of 25 contains 24 unstained slides and 1 stained slide.
Customers Who Bought This Also Bought