Microscope Slides, IHC, SuperWhite Glass, Hydrophilic, 90° Corners
Fournisseur: Mortech Manufacturing
New Product
Certificats
A propos de cet article
Charged SuperWhite glass microscope slides for use in immunohistochemistry.
- Compatible with most auto-stainers, including Ventana
- Standard size 75×25 mm, with 1 mm thickness
- Hydrophilic glass withstands extreme staining environments
- Strong electrostatically charged surface
- Pure white, ultra-clear glass
Slides are available in white, blue, and pink. The packaging options include 1 box of 72 slides and 1 case of 1440 slides.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.