Microscope Slides, Charged, 45° Corners
Fournisseur: Mortech Manufacturing
New Product
Certificats
A propos de cet article
High-quality, charged microscope slides with color coding.
- 1×3" slides, 1 mm thickness
- 100% guaranteed against sticking and contamination
- 45 degree corners
- Uniform flat paint for perfect barcodes
Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac.
Packaging: The packaging options include 1 box of 72 slides and 1 case of 1440 slides.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.