Density Cube, Tin (Sn)
# de catalogue 470345-164
Fournisseur: Wards
Spécifications
- Description:Density cube, Tin (Sn)
- Cat. no.:470345-164
Spécifications
A propos de cet article
Made from tin with an element stamp, measures 2 cm(0.8") per side, perfect for density, specific gravity, and specific heat activities in classrooms and labs.
- Single, solid tin density cube, with element stamp (Sn) for easy identification
- Measures 2 cm cubed
- Excellent for physics classrooms
- Density cubes have a wide range of educational applications for all grade levels, including density investigation, specific heat and specific gravity exploration
The Eisco labs density investigation series is designed to help students identify and figure out mass, understand the relationship between mass/volume/density, understand specific heat and specific gravity, and other introductory physics and chemistry concepts.
Caution: Due to the nature of raw metals, some oxidation or corrosion due to humidity and storage may occur. Simply scuff any oxidation off to make cube look brand new again. Designed to be used in an educational setting and are not fabricated using ultra high precision machining. Expect tolerances in the ±5% range of any stated dimensions (although they are generally much tighter).
Customers Who Bought This Also Bought