Float and Spin Magnet Demonstrator
# de catalogue 470352-576
Fournisseur: United Scientific Supplies
New Product
Spécifications
- Description:Float and spin magnet demonstrator
- Cat. no.:470352-576
- Dimensions:2¹/₂×5×3”
Spécifications
A propos de cet article
Demonstrate the wonders of magnetic levitation.
- Smooth rotation
- No batteries required
This attention-getting demonstration of magnetic repulsion uses a clear acrylic plate that keeps a spinning axle oriented above magnets built into the base of the apparatus. With a simple twist of your fingers, you can start this levitating magnet spinning and kick start a classroom discussion. The black and white coloring makes the demonstration easy to see and helps track the speed of the rotation. The axle measures 5” long.
Customers Who Bought This Also Bought