Dissecting Trays without Wax
Fournisseur: United Scientific Supplies
New Product
Certificats
A propos de cet article
Dissection tray with tie down holes.
- Aluminum construction
- Rounded corners include tie-down holes
- Wax holds dissection pins securely
Dissecting pans are made of durable aluminum with rounded corners. The corners include holes that can be used as tie-down points to hold specimens securely during dissection.
Customers Who Bought This Also Bought