ReadiLink™ Cy5 Nick Translation dsDNA Labeling Kit
Fournisseur: AAT BIOQUEST INC
Certificats
A propos de cet article
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
- Provides all necessary reagents
- Adjustable ratios
- Wide sample type compatibility
The resulted Cy5-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridization (FISH).
Packaging: Packaged in plastic vials.
Caution: For research use only.
Customers Who Bought This Also Bought