ReadiLink™ Biotin Oligo and ssDNA Labeling Kit
Fournisseur: AAT BIOQUEST INC
Certificats
A propos de cet article
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Efficient labelling
- Contains all essential reagents
- Multiple applications
The resulting biotinylated oligo and ssDNA probes are ideally suited for a variety of biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.
Customers Who Bought This Also Bought