ReadiLink™ Cy5 Oligo and ssDNA Labeling Kit
Fournisseur: AAT BIOQUEST INC
Certificats
A propos de cet article
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Robust protocol
- Bright labeling
The resulting Cy5-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.
Customers Who Bought This Also Bought