ReadiLink™ iFluor® 647 FISH Fluorescence Imaging Kit
# de catalogue 76779-036
Fournisseur: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:
Spécifications
- Description:Convenient tool for labeling a target DNA using an iFluor® 647 labeled FISH probe via in situ hybridization, for PCR microplates
- Size:25 reactions
Spécifications
A propos de cet article
The kit provides Taq DNA polymerase enzyme which incorporates iFluor® 647-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
- Robust protocol
- Bright labeling
- Photostable fluorescence
The Readilink™ iFluor® 647 FISH fluorescence imaging kit is a convenient tool for labeling a target DNA using an iFluor® 647 labeled FISH probe via in situ hybridization.
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
Packaging: Packaged in plastic vials.
Caution: For research use only.
Customers Who Bought This Also Bought