AF532 PEG4 biotin conjugate
# de catalogue 77007-312
Fournisseur: AAT BIOQUEST INC
New Product
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:1 mg
Spécifications
Résultats des tests de spécification
- AF532 PEG4 biotin conjugate:
Résultats des tests de spécification
A propos de cet article
:AF532 PEG4 biotin conjugate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Point d'ébullition:
:Boiling Pt:
:Point de fusion:
:Point eclaire:
:Flash Pt:
:M.W. 1172,48 g/mol
MW:1172.48 g/mol
Customers Who Bought This Also Bought