PIP® Corded Ear Plugs
Catalog # 76944-646
Supplier: PIP
Specifications
- NRR:27 dB
- Description:Corded ear plugs
- Cat. no.:76944-646
Specifications
About this item
Reusable TPR corded ear plugs provide NRR 27dB protection.
- Pre-formed plug shape with no rolling required makes insertion simple
- Self-sealing flanges give all day comfort
- Rigid stem makes insertion easy and reduces the risk of contamination
- Non-resonating vinyl cord for comfort and easy storage
- Includes removable cord
Self-sealing, 4-flange TPR (thermoplastic rubber) construction for all-day use and secure sound attenuation. The TPR plug can be cleaned using soap and water, and can be reused several times, making them very cost-efficient. Moderate attenuation allows for safe usage that blocks out hazardous noise yet allows warning signals and voices to be clearly understood.
Customers Who Bought This Also Bought