PIP® Bouton® Universal Fit Polycarbonate Safety Visor with Chin Cup
# de catalogue 76944-634
Fournisseur: PIP
New Product
Spécifications
- Description:Universal fit safety visor with chin cup
- Material:Polycarbonate
- Thickness:0.040"
Spécifications
A propos de cet article
Universal fit polycarbonate safety visor with chin cup features an 0.040" thick lens.
- Clear polycarbonate visor
- Pre-assembled clear chin cup provides additional protection
- Universal hole pattern will fit most headgear and hard hat adapters
- Ships with masking on both sides to prevent scratching
Polycarbonate safety visor with aluminum binding is ideal for high impact applications. Outperforms PETG (Polyethylene Terephthalate Glycol) in a wider range of temperatures.
Certifications: Meets ANSI Z87.1+ requirements.
Customers Who Bought This Also Bought