Spécifications
- No. of Tests:200 Tests
Spécifications
A propos de cet article
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
- FITC filter set compatible
- bright fluorescence
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is imaged with a FITC filter set. Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay