ContactUsLinkComponent
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
# de catalogue 76779-104
Fournisseur:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
PROLIFERATION KIT BUCCULITE XDU CELL
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
# de catalogue 76779-104
Fournisseur:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • No. of Tests:
    200 Tests
Spécifications
A propos de cet article
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • FITC filter set compatible
  • bright fluorescence
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is imaged with a FITC filter set. Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay
Customers Who Bought This Also Bought