Spécifications
- No. of Tests:100 Tests
Spécifications
A propos de cet article
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
- violet laser compatible
- highly reliable
- optimized for flow cytometry
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with mFluor™ Violet 450 MTA. The resulted mFluor™ Violet 450-labeled DNA formed in cells is visualized in Pacific Blue Channel. Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay