Spécifications
- Cat. no.:76779-092
- No. of Tests:200 Tests
Spécifications
A propos de cet article
The kit does not require cell lysis, long incubations or removal of cells with minimal hands-on time. It is compatible with high throughput screening.
- bright fluorescence
- green color
- improved accuracy
The kit uses our Nuclear Green LCS1, a proprietary fluorescent dye that exhibits strong fluorescence upon binding to nucleic acids. The use of Nuclear Green LCS1 improves accuracy over cell metabolism-based cell proliferation or cytotoxicity assays that can be influenced by cell changes that are unrelated to cell numbers.
Sensitive fluorescence-based method for quantifying cells and assessing cell proliferation and cytotoxicity in a microplate format