ContactUsLinkComponent
Cell Meter™ Cell Proliferation Assay Kit
Cell Meter™ Cell Proliferation Assay Kit
# de catalogue 76779-092
Fournisseur:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
PROLIFERATION KIT CELL METER CELL
Cell Meter™ Cell Proliferation Assay Kit
# de catalogue 76779-092
Fournisseur:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    76779-092
  • No. of Tests:
    200 Tests
Spécifications
A propos de cet article
The kit does not require cell lysis, long incubations or removal of cells with minimal hands-on time. It is compatible with high throughput screening.
  • bright fluorescence
  • green color
  • improved accuracy
The kit uses our Nuclear Green LCS1, a proprietary fluorescent dye that exhibits strong fluorescence upon binding to nucleic acids. The use of Nuclear Green LCS1 improves accuracy over cell metabolism-based cell proliferation or cytotoxicity assays that can be influenced by cell changes that are unrelated to cell numbers.
Sensitive fluorescence-based method for quantifying cells and assessing cell proliferation and cytotoxicity in a microplate format
Customers Who Bought This Also Bought