ContactUsLinkComponent
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
# de catalogue 76779-110
Fournisseur:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
PROLIFERATION KIT BUCCULITE XDU CELL
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
# de catalogue 76779-110
Fournisseur:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • No. of Tests:
    200 Tests
Spécifications
A propos de cet article
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay
  • alternative to anti-BrdU
  • extremely sensitive
  • deep red fluorescence
The resulted iFluor® 647-labeled DNA formed in cells is imaged with a Cy5 filter set.
Uses XdU which is incorporated into cellular DNA during DNA synthesis
Customers Who Bought This Also Bought