Spécifications
- Description:DNA/chromosome staining lab activity
- Cat. no.:470336-172
- Includes:SDS and instructions
- No. of Uses:5
Spécifications
A propos de cet article
Prepare stained slides to view plant samples in various stages of development.
- Teacher's manual and study guide are included
Prepare your own squashed stained slide and be able to identify the phases of plant mitosis and chromosomal development. Students will be able to prepare, stain and mount slides using specially prepared onion root tips. Kit contains enough material for 15 groups. Teacher’s Manual and Student Study Guide copymasters are included.