Spécifications
- Description:Introduction to streak isolation and aseptic technique
- Cat. no.:470336-154
- Includes:SDS and instructions
- No. of Uses:10
Spécifications
A propos de cet article
Indroduce skills that microbiologists use on a daily basis.
- Learn about different types of media and how bacteria reproduce
In this activity, students will be introduced to the skills and techniques that microbiologists use on a daily basis. Students will practice these techniques by performing a streak isolation with the provided bacterial cultures, as well as samples they collect from their classroom, to learn about different types of media and how bacterium reproduce.
10 Sterile cotton swabs, pack/2.