ContactUsLinkComponent
Wards® Red Cabbage Juice pH Indicator
Wards® Red Cabbage Juice pH Indicator
# de catalogue 470334-898
Fournisseur:  Ward's Science
RED CABBAGE JUICE KIT
Wards® Red Cabbage Juice pH Indicator
# de catalogue 470334-898
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Red cabbage juice pH indicator
  • Cat. no.:
    470334-898
  • Includes:
    SDS and instructions
Spécifications
A propos de cet article
A colorful way to test the pH of various solutions!
  • Comes with 20 g of red cabbage powder and instructions
Learn about acids, bases, and pH indicators with our red cabbage juice kit. A pH indicator is a chemical that behaves differently depending on the pH level it is exposed to. Red cabbage juice is a natural pH indicator that changes color from red to purple to yellow-green as the pH of a solution increases. Test out different substances to determine if they are acidic or basic, and see if you can make all six colors found on the color chart!
Delivery: 20g Red cabbage powder.
Customers Who Bought This Also Bought