Accessories for OV 625 Digital Disperser
# de catalogue 76416-174
Fournisseur: VELP SCIENTIFIC INC.
Spécifications
- Description:Double rod mini H-stand set
- Cat. no.:76416-174
Spécifications
A propos de cet article
The Double rod mini H-stand set is designed for light weigh instruments and comes complete of all its parts. This stand is designed to avoid rotation of attachment and instruments thanks to the double rod and the bosshead clamp.
Customers Who Bought This Also Bought