OPTI-LYNX™ Holder Cap
# de catalogue 77100-362
Fournisseur: Optimize
Spécifications
- Description:OPTI-LYNX™ holder cap
- Cat. no.:77100-362
- Supplier No.:11-02333-AA
Spécifications
A propos de cet article
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
- For use with OPTI-LYNX™ hardware
- Accepts OPTI-LYNX™ holder tubes of any length
- Connects to ¹/₁₆" outer Ø. tubing with a 10-32 fitting
- A necessary part of every OPTI-LYNX™ configuration
Switching between modes of pre-column protection is as easy as opening OPTI-LYNX™ hardware. Simply select the correct holder tube for the OPTI-LYNX™ insert you wish to use (a guard column, pre-column filter, or straight through connection without protection) thread it into the holder cap, pop in your insert, you're ready to go.
Customers Who Bought This Also Bought