OPTI-LYNX™ Direct-Connect Union Kit
# de catalogue 77100-310
Fournisseur: Optimize
Spécifications
- Description:OPTI-LYNX™ Direct-connect union kit
- Cat. no.:77100-310
- Supplier No.:11-02226-AB
Spécifications
A propos de cet article
If you are already using our OPTI-LYNX™ Guard Column/Pre-Column Filter Kits and want to use OPTI-LYNX™ at the bottom of the column as well, upgrade with this kit.
- Quick-connect union solution
- Quarter-turn design rated to 6,000 psi
- Low Volume/ Low Dispersion interface for volume critical regions
- True zero-dead-volume connection
- Hardware is only plumbed once
- Hand-tight - no tools required
It includes everything you need to equip one end of any analytical column with straight-through quick-connect operation. Can be used alone or in conjunction with other OPTI-LYNX™ products.
Delivery: Includes: Universal port adapter, holder cap, sealing element tube, sealing element and fitting.
Customers Who Bought This Also Bought