EXP® Nano Fittings for 6-40 Ports and ¹/₃₂" Stainless Steel Tubing
Fournisseur: Optimize
Certificats
A propos de cet article
The EXP Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between ¹/₃₂" or ¹/₁₆" tubing and any 6-32, 6-40 or 10-32 port.
- Hand-Tight - rated to 20000+ psi (1400+ bar), tested to 30000 psi
- Auto-adjusting ZDV Connection
- Intended for many repeat uses
- No more cut-off and replacement of swaged ferrules
- Cost efficient with replaceable components
- Simple and versatile - can be used in UHPLC and HPLC applications
The patented Titanium Hybrid Ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The Hybrid Ferrule provides a perfect seal with every connection. The swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides value through repeated zero-dead-volume connections with any port configuration.
Ordering information: Available in standard (¹/₁₆") and nano (¹/₃₂") configurations.
Customers Who Bought This Also Bought