Order Entry
ContactUsLinkComponent
Anti-AAV2 Rabbit Monoclonal Antibody [clone: 15G4]
Anti-AAV2 Rabbit Monoclonal Antibody [clone: 15G4]
# de catalogue 77007-532
Fournisseur:  Genscript
New Product
Anti-AAV2 Rabbit Monoclonal Antibody [clone: 15G4]
# de catalogue 77007-532
Fournisseur:  Genscript
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antigen name:
    AAV2
  • Clonality:
    Monoclonal
  • Clone:
    15G4
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    Lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    It is stable up to 1 year at −20 °C from date of receipt. The reconstituted antibody can be stored for 2 - 3 weeks at 2...8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
  • Immunogen:
    AAV2 Virus-Like Particles (VLPs)
  • Tested applications:
    Dot blot
  • Cat. no.:
    77007-532
  • Packaging:
    Vial

Spécifications

A propos de cet article

Adeno-associated virus type 2 (AAV2) is a valuable vector for gene therapy. It is one member of the human parvovirus family which depends on co-infection with helper viruses such as adenovirus or herpes virus for efficient reproduction. AAV2 empty capsids are composed of three proteins, VP1, VP2 and VP3 with relative molecular masses of 87, 72 and 62 kDa, respectively.

  • High specificity
  • High affinity
  • High stability

GenScript’s AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen: AAV2
Clonality: Monoclonal
Clone: 15G4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Customers Who Bought This Also Bought