To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antigen name:VHH
- Clonality:Monoclonal
- Clone:45H8
- Host:Rabbit
- Isotype:IgG
- Conjugation:iFluor® 647
- Flow cytometry:Yes
- Size:200 μl
- Cross adsorption:It has no cross-reactivity with mouse, rat, rabbit, or goat immunoglobulins
- Form:Liquid
- Storage buffer:Supplied in PBS (pH 7.2), containing 10 mg/ml BSA, 0.02% sodium azide and 50% glycerol
- Storage temperature:Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Concentration:0.5 mg/ml
- Immunogen:VHH antibody
- Cat. no.:77007-504
- Packaging:Vial
Spécifications
A propos de cet article
A camelid single-domain antibody (sdAb, called Nanobody by Ablynx, the developer) is a peptide chain of about 110 amino acids long, comprising one variable domain of a heavy-chain antibody, these are called VHH fragments. Like a whole antibody, it is able to bind selectively to a specific antigen. Single-domain antibodies allow a broad range of applications in biotechnical as well as therapeutic uses due to their small size, simple production, and high affinity. However, their sequences may increase the risk of immunogenicity and anti-drug antibody (ADA) development in humans, and thus, sdAbs are routinely humanized during development.
- MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
- No cross reactivity with other species
- Specific to camelid IgG2 and IgG3
- Recognize conformational epitopes of the VHH domain
- Recognize both camelid and humanized VHHs
GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.
Type:
Antigen: VHH
Clonality: Monoclonal
Clone: 45H8
Conjugation: iFluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: