Order Entry
ContactUsLinkComponent
Anti-Protein A Mouse Monoclonal Antibody [clone: 4F9A8]
Anti-Protein A Mouse Monoclonal Antibody [clone: 4F9A8]
# de catalogue 77007-544
Fournisseur:  Genscript
New Product
Anti-Protein A Mouse Monoclonal Antibody [clone: 4F9A8]
# de catalogue 77007-544
Fournisseur:  Genscript
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein A
  • Clonality:
    Monoclonal
  • Clone:
    4F9A8
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    40 μg
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    Supplied in PBS, pH 7.4 with 0.02% sodium azide
  • Storage temperature:
    The product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2...8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized antibody with deionized water (or equivalent) to the final concentration of 0.5 mg/ml.
  • Immunogen:
    Purified recombinant Protein A
  • Cat. no.:
    77007-544
  • Packaging:
    Vial

Spécifications

A propos de cet article

Protein A, a bacterial cell wall protein isolated from Staphylococcus aureus, binds to mammalian IgGs mainly through Fc regions. The Protein A is most commonly bound to a chromatography resin, used for production of antibody drugs or reagents. Protein A antibody can be used for quantification of Protein A in drugs and foods or for evaluation of Protein A resin. This Protein A mAb is advantageous due to its IgG1 isotype, for mouse IgG1 has weak binding capacity to Protein A. GenScript Protein A Antibody, mAb, Mouse is produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes obtained from mouse immunized with purified recombinant Protein A.

  • High specificity
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: Protein A
Clonality: Monoclonal
Clone: 4F9A8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Customers Who Bought This Also Bought