Order Entry
ContactUsLinkComponent
Anti-Palivizumab Mouse Monoclonal Antibody [clone: 7B3]
Anti-Palivizumab Mouse Monoclonal Antibody [clone: 7B3]
# de catalogue 77007-474
Fournisseur:  Genscript
New Product
Anti-Palivizumab Mouse Monoclonal Antibody [clone: 7B3]
# de catalogue 77007-474
Fournisseur:  Genscript
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antigen name:
    Palivizumab
  • Clonality:
    Monoclonal
  • Clone:
    7B3
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    40 μg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    This product is lyophilized with PBS, pH 7.2, containing 0.02% sodium azide
  • Storage temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Immunogen:
    Palivizumab
  • Cat. no.:
    77007-474
  • Packaging:
    Vial

Spécifications

A propos de cet article

Palivizumab with trade name Synagis, is an FDA-approved drug for the prevention of respiratory syncytial virus (RSV) infections. It is a recombinant humanized IgG1 kappa isotype monoclonal antibody. Palivizumab binds the RSV envelope fusion protein (RSV F) on the surface of the virus and blocking a critical step in the membrane fusion process. Anti-Palivizumab Antibody (7B3), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Palivizumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Palivizumab
Clonality: Monoclonal
Clone: 7B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Customers Who Bought This Also Bought